Learn More
Cayman Chemical 28-AcetylbetulIn 10mg
SDP
Catalog No. 502948013
Encompass_Preferred
A lupane triterpenoid with anti-inflammatory and anticancer activities; inhibits LPS-induced NO and PGE2 production by 55.9 and 69.5%, respectively, in RAW 264.7 cells (IC50s = 4.7 and 1.7 µM, respectively); induces cytotoxicity in a variety of cancer cells, including A549, HT-29, and MCF-7 cells (IC50s = 14.37, 10.96, and 11.38 µM, respectively)
Catalog No. 50-294-8013 Supplier Cayman Chemical Supplier No. 3305210 MG
A lupane triterpenoid with anti-inflammatory and anticancer activities; inhibits LPS-induced NO and PGE2 production by 55.9 and 69.5%, respectively, in RAW 264.7 cells (IC50s = 4.7 and 1.7 µM, respectively); induces cytotoxicity in a variety of cancer cells, including A549, HT-29, and MCF-7 cells (IC50s = 14.37, 10.96, and 11.38 µM, respectively)