Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ 28# Latex Free Cleanroom Paper, 3-Hole-Drilled

Catalog No. 18003108 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Blue
Canary
Gray
Green
Pink
White
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
18-003-108 Green
18-003-100 Gray
18-003-101 Pink
18-003-102 White
18-003-106 Blue
18-003-107 Canary
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-003-108
Add to Cart
Add to Cart

3-hole-drilled 28# cleanroom paper for controlled environments. Pre-punched for use in cleanroom 3-ring binders.

  • 22#, 3-hole-drilled latex-free cleanroom paper, intended for use in controlled environments.
  • Our low-density polyethylene and latex-free means our cleanroom paper is not limited and is safe to use in almost all controlled environments.
  • Both our 22# and 28# latex-free cleanroom paper have been injected with a low-density polyethylene synthetic copolymer to substantially reduce particle generation.
  • Both have exceptional thickness and strength, with excellent heat resistance. Ideal for use in both laser and inkjet printers.
  • Class 10 or ISO 3 certified.
  • Made in the USA.

Specifications

Material Paper
Color Green
Product Type Cleanroom Paper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.