Learn More
Cayman Chemical 28-(Poc-amino)betulin, Purity: ≥95%, 10 mg
SDP
Catalog No. 502946792
Encompass_Preferred
An alkyne derivative of betulin; can be used for click chemistry applications
Catalog No. 50-294-6792 Supplier Cayman Chemical Supplier No. 3306910 MG
An alkyne derivative of betulin; can be used for click chemistry applications