Learn More
Cayman Chemical 28-(Poc-amino)betulin, Purity: ≥95%, 50 mg
SDP
Catalog No. 502949861
Encompass_Preferred
An alkyne derivative of betulin; can be used for click chemistry applications
Catalog No. 50-294-9861 Supplier Cayman Chemical Supplier No. 3306950 MG
An alkyne derivative of betulin; can be used for click chemistry applications