Learn More
Chemglass Life Sciences 28 x 70 Vials, Amber, 30mL Capacity, 24-400 Thread with PTFE Lined Solid PTFE Caps, Sold in a LAB-PAC of 144pcs
SDP
Catalog No. 50873526
Encompass_Preferred
28 x 70 Vials, Amber, 30mL Capacity, 24-400 Thread with Solid PTFE Caps, Sold In A LAB-PAC of 144 pcs. Amber sample vials packaged in convenient LAB-PACS of 144 pieces. Made from Type I borosilicated glass. Full height partitions minimize breakage. Solid PTFE lined caps. Open top caps with septa are available under part number CG-4910.
Catalog No. 50-873-526 Supplier Chemglass Life Sciences Supplier No. CG490505
28 x 70 Vials, Amber, 30mL Capacity, 24-400 Thread with Solid PTFE Caps, Sold In A LAB-PAC of 144 pcs. Amber sample vials packaged in convenient LAB-PACS of 144 pieces. Made from Type I borosilicated glass. Full height partitions minimize breakage. Solid PTFE lined caps. Open top caps with septa are available under part number CG-4910.