Learn More
Thermo Scientific Dionex AutoTrace 280 Plunger Assemblies | | 1 EA
Catalog No. NC2184051 Shop All Thermo Scientific Products
Encompass
Replacement plunger assemblies for the Dionex AutoTrace 280 Automated SPE system allow you to convert to a different size cartridge
Catalog No. NC2184051 Supplier Thermo Scientific Supplier No. 071080
Replacement plunger assemblies for the Dionex AutoTrace 280 Automated SPE system allow you to convert to a different size cartridge