Learn More
Chemglass Life Sciences Flask, Fernbach, 2800mL, Plain 59mm ID Top, with 3 Bottom Baffles, Approx OD x Height (mm): 210 x 230
SDP
Catalog No. 501215175
Encompass_Preferred
Flask, Fernbach, 2800mL, Plain 59mm ID Top, with 3 Bottom Baffles, Approx OD x Height (mm): 210 x 230. Flasks are designed for culturing organisms that require a large surface area to volume ratio. Flask has 3 baffles on the bottom surface to help oxygen transfer to the culture medium.
Catalog No. 50-121-5175 Supplier Chemglass Life Sciences Supplier No. CLS202415
Flask, Fernbach, 2800mL, Plain 59mm ID Top, with 3 Bottom Baffles, Approx OD x Height (mm): 210 x 230. Flasks are designed for culturing organisms that require a large surface area to volume ratio. Flask has 3 baffles on the bottom surface to help oxygen transfer to the culture medium.