Learn More
Aviva Systems Biology Corp 2810405K02Rik Antibody - middle region (ARP52920_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52920_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against 2810405K02Rik. It was validated on Western Blot. . Alternate names: Fam213b, PM/PGFS, AI836168, RP23-15L19.6, 2810405K02RikApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-285-784