Learn More
Cayman Chemical 28Deoxybetuln mthylenamIn 10mg
SDP
Catalog No. 502949619
Encompass_Preferred
A derivative of betulin; has been used in the synthesis of triterpene derivatives with antiviral activity
Catalog No. 50-294-9619 Supplier Cayman Chemical Supplier No. 3306010 MG
A derivative of betulin; has been used in the synthesis of triterpene derivatives with antiviral activity