Learn More
Braintree Scientific 28g Mouse Intravascular Catheter (sterile)
SDP
Catalog No. 501955029
Encompass
Designed for quick and easy insertion into arteries and veins of mice and rats (primarily mice). 60mm L, 0.014" od x 0.007" id (supplied sterile) **Sold for research purposes only and not for human use**
Catalog No. 50-195-5029 Supplier Braintree Scientific Supplier No. MC0189E0
Designed for quick and easy insertion into arteries and veins of mice and rats (primarily mice). 60mm L, 0.014" od x 0.007" id (supplied sterile) **Sold for research purposes only and not for human use**