Learn More
SAI Infusion Technologies Mouse Femoral Vein Catheter - 17cm PU
SDP
Catalog No. NC0474961
Encompass
Femoral Vein Catheter for Mice under 25g. Polyurethane, 20mm 1Fr tip, 2 suture beads, 17cm total length. Access with 25G blunt needle (such as B25-50). Sterile. Catheters may be used for sampling or infusion. Product is for laboratory animal research only.
Catalog No. NC0474961 Supplier SAI Infusion Technologies Supplier No. MFV02
Femoral Vein Catheter for Mice under 25g. Polyurethane, 20mm 1Fr tip, 2 suture beads, 17cm total length. Access with 25G blunt needle (such as B25-50). Sterile. Catheters may be used for sampling or infusion. Product is for laboratory animal research only.