Learn More
VivaGuard™ 28G Safety Lancets
Catalog No. 23111434
Change view
Click to view available options
Quantity:
100 Lancets/Box
Catalog No. Quantity
23-111-434 100 Lancets/Box
1 options
This item is not returnable. View return policy
Catalog No. 23-111-434 Supplier VivaGuard™ Supplier No. VGL02382
Add to Cart
Add to Cart
This item is not returnable. View return policy
Extra-sharpened, 28G safety lancets are sterile with an anti-skid body and easy twist off, pull, place and press process.
  • Ergonomically designed Safety Lancets for individual use
  • Easy to carry with anti-skid body design to ensure safe grip
  • Easy to use: just twist and pull off the protective cap, place it on the puncture site and press to collect blood sample 28G
  • Sterile Safety Lancets with extra sharpened needlepoints to minimize discomfort
  • Defined penetration depth at 1.8 mm for optimal sampling
  • Well controlled small penetration area for precision
  • Needle automatically retracts and locks, to eliminate re-use and cross infection for ensured safety Convenient storage with 5-year shelf life
  • Additional gauges available
Specifications
Needle Gauge 28G
Depth (Metric) Needle 1.8 mm
Sterility Sterile
Disposable Single-use
For Use With (Equipment) Germaine™ Lancing Device
Product Type Safety Lancet
For Use With (Application) Fingerstick Sample Tests, Capillary Testing
Includes 100 Safety Lancets, Instructions
Quantity 100 Lancets/Box
Depth (English) Needle 0.071 in.