Learn More
PATHOLOGY INNOVATIONS 28MM SMALL CHUCK
Catalog No. NC0754411
Encompass_Preferred
The stainless steel chuck 28mm by 28mm and are designed to be used cold for rapid freezing, and to maximize the gripping power required to hold the embedded tissue block. Chucks can also be used warm with use of the “over-chuck freezing blocks”. These chucks were designed by professor and author of frozen section literature Dr. Stephen Peters to fit most cryostats.
Catalog No. NC0754411 Supplier PATHOLOGY INNOVATIONS Supplier No. 131
The stainless steel chuck 28mm by 28mm and are designed to be used cold for rapid freezing, and to maximize the gripping power required to hold the embedded tissue block. Chucks can also be used warm with use of the “over-chuck freezing blocks”. These chucks were designed by professor and author of frozen section literature Dr. Stephen Peters to fit most cryostats.