Learn More
USA Scientific Inc 28-place upright freezer rack
Catalog No. 502744766
Encompass
28-place (4 long x 7 high) upright freezer rack for 2" H boxes, 21 5/8" L x 5 1/2" W x 15 3/10" H, stainless steel
Catalog No. 50-274-4766 Supplier USA Scientific Inc Supplier No. 26024700
28-place (4 long x 7 high) upright freezer rack for 2" H boxes, 21 5/8" L x 5 1/2" W x 15 3/10" H, stainless steel