Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ 28S rRNA (+) Control CISH Probe

Catalog No. 89335570 Shop All Abnova Corporation Products
Change view
Click to view available options
For Use With (Application):
CISH-P
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
89-335-570 CISH-P
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-570 Supplier Abnova™ Supplier No. CO0001
Add to Cart
Add to Cart

28S rRNA (+) Control CISH Probe is designed for the qualitative detection of human 28S ribosomal RNA in formalin-fixed, paraffin-embedded tissue or cells by chromogenic in situ hybridization (CISH).

The probe is intended to be used in combination with the CISH Implementation HRP-DAB Kit, which provides necessary reagents for specimen pretreatment and post-hybridization processing.

Depending on the detection system that is used, colored precipitates, which can be clearly distinguished from the background, will be observed within the cells targeted by this probe. A positive reactivity for 28S rRNAs in the target cells is indicated by cytoplasmic staining. Visualization of signals should be performed by light microscopy using a 10x or 20x objective. For signal evaluation, necrotic, degenerated or over-digested cells should be avoided as these cells often stain nonspecifically. In order to judge the specificity of the hybridization signals and to confirm the correct performance of the method, any hybridization should be accompanied by controls. We recommend using at least one control sample containing both true positive and negative staining cells.

Specifications

Content And Storage Bring probe to hybridization temperature before use.
For Use With (Application) CISH-P
Includes Reagent Provided:

Digoxigenin-labeled oligonucleotides targeting 28S ribosomal RNA.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.