Learn More
Ace Glass, Inc. Complete 29/42 10mm bearing, straight adapter style
SDP
Catalog No. 509558178
Encompass_Preferred
29/42 10mm straight-adapter style bearing includes all parts needed. Self-aligning, lubricant free PTFE Trubore bearing is designed for use with 10mm O.D. stainless steel or polished glass shafts.
Catalog No. 50-955-8178 Supplier Ace Glass, Inc. Supplier No. 806646
29/42 10mm straight-adapter style bearing includes all parts needed. Self-aligning, lubricant free PTFE Trubore bearing is designed for use with 10mm O.D. stainless steel or polished glass shafts.