Learn More
Ace Glass, Inc. 29/42 10mm debris trap, glass only
SDP
Catalog No. 50975972
Encompass_Preferred
29/42 10mm glass portion for PTFE Trubore debris trap. Use with ACE pstainless steel or polished glass 10mm O.D. stir shafts.
Catalog No. 50-975-972 Supplier Ace Glass, Inc. Supplier No. 8066224
29/42 10mm glass portion for PTFE Trubore debris trap. Use with ACE pstainless steel or polished glass 10mm O.D. stir shafts.