Learn More
Ace Glass, Inc. 29/42 10mm straight adapter, hose connection port, glass only
SDP
Catalog No. 50975962
Encompass_Preferred
29/42 10mm straight adapter glass portion only, with hose connection port, for use with self-aligning, lubricant free PTFE Trubore bearing or combination bearing / debris trap. Use with 10mm O.D. stainless steel or polished glass shafts.
Catalog No. 50-975-962 Supplier Ace Glass, Inc. Supplier No. 806671
29/42 10mm straight adapter glass portion only, with hose connection port, for use with self-aligning, lubricant free PTFE Trubore bearing or combination bearing / debris trap. Use with 10mm O.D. stainless steel or polished glass shafts.