Learn More
Ace Glass, Inc. Complete 29/42 10mm bearing, straight adapter style with hose connection port
SDP
Catalog No. 50979644
Encompass_Preferred
29/42 10mm self-aligning, lubricant-free PTFE Trubore straight adapter style bearing with hose connection port. Use with ACE polished stainless steel or glass 10mm O.D. stir shafts. Can achieve vacuum down to 1mm Hg. Multi-directional.
Catalog No. 50-979-644 Supplier Ace Glass, Inc. Supplier No. 8066132
29/42 10mm self-aligning, lubricant-free PTFE Trubore straight adapter style bearing with hose connection port. Use with ACE polished stainless steel or glass 10mm O.D. stir shafts. Can achieve vacuum down to 1mm Hg. Multi-directional.