Learn More
Ace Glass, Inc. Complete 29/42 10mm bearing, debris trap style
SDP
Catalog No. 50975976
Encompass_Preferred
29/42 10mm self-aligning, lubricant free PTFE Trubore debris-trap style bearing includes all parts needed. Use with ACE 10mm O.D. stainless steel or polished glass shafts. Bearing is multi-directional, and achieves vacuum down to 1mm Hg.
Catalog No. 50-975-976 Supplier Ace Glass, Inc. Supplier No. 8066324
29/42 10mm self-aligning, lubricant free PTFE Trubore debris-trap style bearing includes all parts needed. Use with ACE 10mm O.D. stainless steel or polished glass shafts. Bearing is multi-directional, and achieves vacuum down to 1mm Hg.