Learn More
Ace Glass, Inc. 29/42 2000ML BOTTLE 29/42 2000ML BOTTLE
SDP
Catalog No. 50971854
Encompass_Preferred
2000 mL single neck bottle made of heavy wall glass, with a reinforced 29/42 standard taper outer joint.
Catalog No. 50-971-854 Supplier Ace Glass, Inc. Supplier No. 534520
2000 mL single neck bottle made of heavy wall glass, with a reinforced 29/42 standard taper outer joint.