Learn More
Ace Glass, Inc. 29/42 20ML MOISTURE RCVR 29/42 20ML MOISTURE RCVR
SDP
Catalog No. 50975348
Encompass_Preferred
20mL moisture receiver with 29/42 joint. Includes a 120¿, 1:5 PTFE stopcock which provides adequate shut-off in the intermediate position. Thermometer joint is 10/30.
Catalog No. 50-975-348 Supplier Ace Glass, Inc. Supplier No. 774704
20mL moisture receiver with 29/42 joint. Includes a 120¿, 1:5 PTFE stopcock which provides adequate shut-off in the intermediate position. Thermometer joint is 10/30.