Learn More
Ace Glass, Inc. Connecting adapter, enlarging, 29/42 outer top, 24/40 inner bottom
SDP
Supplier:  Ace Glass, Inc. 500524
Encompass_Preferred
Connecting, enlarging adapter featuring a 29/42 outer top female joint and a 24/40 inner bottom male joint.
Catalog No. 50-955-6874