Learn More
Ace Glass, Inc. Connecting adapter, reducing, 29/42 outer top, 45/50 inner bottom
SDP
Catalog No. 50971563
Encompass_Preferred
Catalog No. 50-971-563 Supplier Ace Glass, Inc. Supplier No. 500056