Learn More
Ace Glass, Inc. 29/42 61.0cm distillation column
SDP
Catalog No. 50973272
Encompass_Preferred
61.0cm Hemple distillation column with 29/42 standard taper joints, and indentations to support packing. Length in centimeters refers to the length from the packing support to the bottom of the outer ground joint.
Catalog No. 50-973-272 Supplier Ace Glass, Inc. Supplier No. 657214
61.0cm Hemple distillation column with 29/42 standard taper joints, and indentations to support packing. Length in centimeters refers to the length from the packing support to the bottom of the outer ground joint.