Learn More
Ace Glass, Inc. Adapter, 29/42, 3/8in size D hose connection, #11 Ace-Thred complete with 7506-02 nylon bushing and 7855-708 O-ring
SDP
Supplier:  Ace Glass, Inc. 526140
Encompass_Preferred
Adapter with a 29/42 joint at the bottom, 3/8 size D serrated hose connection, and a #11 Ace-Thred at top. Complete with nylon bushing and O-ring.
Catalog No. 50-971-801