Learn More
Ace Glass, Inc. Adapter, 29/42, 3/8in size D hose connection, #15 Ace-Thred, complete with 7506-06 nylon bushing and 7855-716 O-ring
SDP
Catalog No. 50971804
Encompass_Preferred
Adapter with a 29/42 joint at the bottom, 3/8 size D serrated hose connection, and a #15 Ace-Thred at top. Complete with nylon bushing and O-ring.
Catalog No. 50-971-804 Supplier Ace Glass, Inc. Supplier No. 526159
Adapter with a 29/42 joint at the bottom, 3/8 size D serrated hose connection, and a #15 Ace-Thred at top. Complete with nylon bushing and O-ring.