Learn More
Ace Glass, Inc. Adapter, 29/42, 3/8in size D hose connection, #11 Ace-Thred, glass only
SDP
Supplier:  Ace Glass, Inc. 526139
Encompass_Preferred
Adapter with a 29/42 joint at the bottom, 3/8 size D serrated hose connection, and a #11 Ace-Thred at top. Does not include nylon bushing or O-ring.
Catalog No. 50-971-800