Learn More
Ace Glass, Inc. Adapter, hose connection, bent, 29/42
SDP
Catalog No. 509556975
Encompass_Preferred
Adapter with a 3/8-inch 90-degree hose connection top, and an inner 29/42 standard taper joint bottom.
Catalog No. 50-955-6975 Supplier Ace Glass, Inc. Supplier No. 520515
Adapter with a 3/8-inch 90-degree hose connection top, and an inner 29/42 standard taper joint bottom.