Learn More
Ace Glass, Inc. 29/42 ADAPTER GAS INLET 29/42 ADAPTER GAS INLET
SDP
Catalog No. 50971738
Encompass_Preferred
Side tube with hose connection. Stem below joint is 175mm. Hose Conn. size D. Available in 24/40 and 29/42 standard joints or 35/25 spherical joints.
Catalog No. 50-971-738 Supplier Ace Glass, Inc. Supplier No. 519015
Side tube with hose connection. Stem below joint is 175mm. Hose Conn. size D. Available in 24/40 and 29/42 standard joints or 35/25 spherical joints.