Learn More
Chemglass Life Sciences Joint Size: 29/42, Total Length: min. 124 (mm), max. 142 (mm), Bellows Adapter
SDP
Catalog No. 50871885
Encompass_Preferred
Adapter is constructed completely of PTFE and is ideal for eliminating stress between standard joints. Can be bent up to 120¿. Top outer and lower inner are of the same size. Joint Size: 29/42,Total Length: min. 124 (mm), max. 142 (mm)
Catalog No. 50-871-885 Supplier Chemglass Life Sciences Supplier No. CG100421
Adapter is constructed completely of PTFE and is ideal for eliminating stress between standard joints. Can be bent up to 120¿. Top outer and lower inner are of the same size. Joint Size: 29/42,Total Length: min. 124 (mm), max. 142 (mm)