Learn More
Ace Glass, Inc. 29/42 DISTILLING HEAD 29/42 DISTILLING HEAD
SDP
Catalog No. 50978835
Encompass
Magnetically operated swinging funnel type distillation head with iron slug totally enclosed. Includes 10/30 thermometer joint with 76mm immersion, 28/11 side arm joint, 4mm stopcock bore, and 29/42 column joint.
Catalog No. 50-978-835 Supplier Ace Glass, Inc. Supplier No. 659410
Magnetically operated swinging funnel type distillation head with iron slug totally enclosed. Includes 10/30 thermometer joint with 76mm immersion, 28/11 side arm joint, 4mm stopcock bore, and 29/42 column joint.