Learn More
Chemglass Life Sciences 29/42 Inner Joint, Un-Cut Drip, on 28mm OD x 2.0mm W tubing
SDP
Catalog No. CG11715
Encompass_Preferred
29/42 Inner Joint, Un-Cut Drip, on 28mm OD x 2.0mm W tubing. Drip tip. Joints are available uncut or with the drip tip being cut at a 45º angle. Tooled on the same tubing sizes as the CG-100 inner joints.
Catalog No. CG11715 Supplier Chemglass Life Sciences Supplier No. CG11715
29/42 Inner Joint, Un-Cut Drip, on 28mm OD x 2.0mm W tubing. Drip tip. Joints are available uncut or with the drip tip being cut at a 45º angle. Tooled on the same tubing sizes as the CG-100 inner joints.