Learn More
Ace Glass, Inc. 29/42 inner joint, stainless steel, unthreaded
SDP
Catalog No. 50975253
Encompass
29/42 inner joint made of stainless steel. Joint is unthreaded and machined from 18-8 type 303 free machining stainless steel and supplied in accordance with CS21-58 of the NIST.
Catalog No. 50-975-253 Supplier Ace Glass, Inc. Supplier No. 765126
29/42 inner joint made of stainless steel. Joint is unthreaded and machined from 18-8 type 303 free machining stainless steel and supplied in accordance with CS21-58 of the NIST.