Learn More
Chemglass Life Sciences Vapor Tube, Steam Duct, 45mm, 29/42 Joint, Compatible with Buchi Rotary Evaporators, Combi-Clip Not Included, 172mm Approx OAL
SDP
Catalog No. CG131730
Encompass_Preferred
Vapor Tube, Steam Duct, 45mm, 29/42 Joint, Compatible with Buchi Rotary Evaporators, Combi-Clip Not Included, 172mm Approx OAL. Replacement Vapor tubes compatible with Buchi rotary evaporators. Thread is made for use with standard Combi-Clip. Available in the 4 listed sizes. NOT SUPPLIED WITH COMBI-CLIP.Dimension “A”:CG-1317-30: 45mm
Catalog No. CG131730 Supplier Chemglass Life Sciences Supplier No. CG131730
Vapor Tube, Steam Duct, 45mm, 29/42 Joint, Compatible with Buchi Rotary Evaporators, Combi-Clip Not Included, 172mm Approx OAL. Replacement Vapor tubes compatible with Buchi rotary evaporators. Thread is made for use with standard Combi-Clip. Available in the 4 listed sizes. NOT SUPPLIED WITH COMBI-CLIP.Dimension “A”:CG-1317-30: 45mm