Learn More
Chemglass Life Sciences Adapter, Distillation, 105°, 29/42 Joint Size
SDP
Catalog No. CG107102
Encompass_Preferred
Adapter, Distillation, 105¿, 29/42 Joint Size. Distillation adapter with top outer and lower inner joints both being the same size.
Catalog No. CG107102 Supplier Chemglass Life Sciences Supplier No. CG107102
Adapter, Distillation, 105¿, 29/42 Joint Size. Distillation adapter with top outer and lower inner joints both being the same size.