Learn More
Chemglass Life Sciences 29/42 Outer Joint, Dewar Seal, 60mm OD Flare
SDP
Catalog No. CG13135
Encompass_Preferred
29/42 Outer Joint, Dewar Seal, 60mm OD Flare. Used in construction of apparatus requiring the outer member to be jacketed. Configuration of flare permits seal to be made without having to regrind the joint.
Catalog No. CG13135 Supplier Chemglass Life Sciences Supplier No. CG13135
29/42 Outer Joint, Dewar Seal, 60mm OD Flare. Used in construction of apparatus requiring the outer member to be jacketed. Configuration of flare permits seal to be made without having to regrind the joint.