Learn More
Chemglass Life Sciences Chemglass 29/42 Lower Joint Adapter for 10 mm Stir Shaft - Mechanical seal bearing, 1-3/4 in threaded connection (CG-2077-M-38)
SDP
Catalog No. NC2094956
Encompass_Preferred
Lower joint adapter for 29/42 ground joints that provides a dry-running mechanical seal for 10 mm stir shafts and a 1-3/4 in threaded connection. Supplied complete with perfluorinated O-rings and a loosening nut, the adapter resists aggressive chemicals while minimizing contamination and allowing vertical adjustment for different agitator styles.
  • Provides a vacuum- and pressure-tight seal for 29/42 lower joints.
  • Designed for 10 mm stir shafts and 1-3/4 in threaded connections.
  • Includes perfluorinated O-rings and a loosening nut for chemical resistance.
  • Dry-running mechanical seal rotates with the shaft to prevent contamination.
  • Rated to 5 Torr vacuum and 2-3 psi pressure; seals up to 2000 rpm.
  • Compatible with glass, metal, or PTFE-covered stir shafts and allows vertical adjustment.
This item is not returnable. View return policy
Catalog No. NC2094956 Supplier Chemglass Life Sciences Supplier No. CG2077M38
Lower joint adapter for 29/42 ground joints that provides a dry-running mechanical seal for 10 mm stir shafts and a 1-3/4 in threaded connection. Supplied complete with perfluorinated O-rings and a loosening nut, the adapter resists aggressive chemicals while minimizing contamination and allowing vertical adjustment for different agitator styles.
  • Provides a vacuum- and pressure-tight seal for 29/42 lower joints.
  • Designed for 10 mm stir shafts and 1-3/4 in threaded connections.
  • Includes perfluorinated O-rings and a loosening nut for chemical resistance.
  • Dry-running mechanical seal rotates with the shaft to prevent contamination.
  • Rated to 5 Torr vacuum and 2-3 psi pressure; seals up to 2000 rpm.
  • Compatible with glass, metal, or PTFE-covered stir shafts and allows vertical adjustment.