Learn More
Chemglass Life Sciences 29/42 Outer Joint, 32mm X 2.4W Tubing
SDP
Catalog No. 501531892
Encompass_Preferred
29/42 Outer Joint, 32mm X 2.4W Tubing. Standard taper outer joint tooled on tubing approximately the same diameter as the top reinforcing ring, affording no hold-up below joint.
Catalog No. 50-153-1892 Supplier Chemglass Life Sciences Supplier No. CGB10107
29/42 Outer Joint, 32mm X 2.4W Tubing. Standard taper outer joint tooled on tubing approximately the same diameter as the top reinforcing ring, affording no hold-up below joint.