Learn More
Ace Glass, Inc. 29/42 straight-sided outer joint. O.D. of standard taper joint and tube are approximately the same size, allowing for ease in clamping.
SDP
Catalog No. 50979340
Encompass_Preferred
29/42 straight-sided outer joint. O.D. of standard taper joint and tube are approximately the same size, allowing for ease in clamping.
Catalog No. 50-979-340 Supplier Ace Glass, Inc. Supplier No. 756738
29/42 straight-sided outer joint. O.D. of standard taper joint and tube are approximately the same size, allowing for ease in clamping.