Learn More
Chemglass Life Sciences 29/42 Outer Joint, Full Length, on 32mm OD x 2.4mm W tubing
SDP
Catalog No. CG10107
Encompass_Preferred
Standard taper outer joint tooled on tubing approximately the same diameter as the top reinforcing ring, affording no hold-up below joint. 29/42 Outer Joint, Full Length, on 32mm OD x 2.4mm W tubing.
Catalog No. CG10107 Supplier Chemglass Life Sciences Supplier No. CG10107
Standard taper outer joint tooled on tubing approximately the same diameter as the top reinforcing ring, affording no hold-up below joint. 29/42 Outer Joint, Full Length, on 32mm OD x 2.4mm W tubing.