Learn More
Chemglass Life Sciences 29/42 Outer Joint, Full Length, on 28mm OD x 2mm W tubing
SDP
Catalog No. CG10207
Encompass_Preferred
Standard taper outer joint tooled on the same size tubing as the CG-100 inner joints. All CG-102 outer joints are supplied standard with rings, but are available tooled without rings on a special order basis. 29/42 Outer Joint, Full Length, on 28mm OD x 2mm W tubing.
Catalog No. CG10207 Supplier Chemglass Life Sciences Supplier No. CG10207
Standard taper outer joint tooled on the same size tubing as the CG-100 inner joints. All CG-102 outer joints are supplied standard with rings, but are available tooled without rings on a special order basis. 29/42 Outer Joint, Full Length, on 28mm OD x 2mm W tubing.