Learn More
Ace Glass, Inc. 29/42 outer joint, stainless steel, unthreaded
SDP
Catalog No. 50975254
Encompass
29/42 outer joint made of stainless steel. Joint is unthreaded and machined from 18-8 type 303 free machining stainless steel and supplied in accordance with CS21-58 of the NIST.
Catalog No. 50-975-254 Supplier Ace Glass, Inc. Supplier No. 765128
29/42 outer joint made of stainless steel. Joint is unthreaded and machined from 18-8 type 303 free machining stainless steel and supplied in accordance with CS21-58 of the NIST.