Learn More
Ace Glass, Inc. 29/42 outer joint, stainless steel, 3/4" male NPT
SDP
Catalog No. 50975257
Encompass
29/42 outer joint made of stainless steel. Joint is machined with male NPT thread, manufactured from 18-8 type 303 free machining stainless steel, and supplied in accordance with CS21-58 of the NIST.
Catalog No. 50-975-257 Supplier Ace Glass, Inc. Supplier No. 7651228
29/42 outer joint made of stainless steel. Joint is machined with male NPT thread, manufactured from 18-8 type 303 free machining stainless steel, and supplied in accordance with CS21-58 of the NIST.