Learn More
Chemglass Life Sciences Sleeve, PTFE, 29/42 Joint Size
SDP
Catalog No. CG13705
Encompass_Preferred
Sleeve, PTFE, 29/42 Joint Size. A .002in thick PTFE sleeve for standard taper joints provides a grease-free seal. Sold in packages of 3 each.
Catalog No. CG13705 Supplier Chemglass Life Sciences Supplier No. CG13705
Sleeve, PTFE, 29/42 Joint Size. A .002in thick PTFE sleeve for standard taper joints provides a grease-free seal. Sold in packages of 3 each.