Learn More
Chemglass Life Sciences Sleeve, PTFE, Full Length, 29/42 Joint Size
SDP
Catalog No. CG11616
Encompass_Preferred
Sleeve, PTFE, Medium Length, 29/42 Joint Size. Reusable .005in thick PTFE sleeve for standard taper joints provides a grease-free seal.
Catalog No. CG11616 Supplier Chemglass Life Sciences Supplier No. CG11616
Sleeve, PTFE, Medium Length, 29/42 Joint Size. Reusable .005in thick PTFE sleeve for standard taper joints provides a grease-free seal.