Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 29-amino-3,6,9,12,15,18,21,24,27-nonaoxanonacosan-1-ol | 129449-09-8 | >98.0% | 457.55 g/mol | C20H43NO10 | 100 MG
SDP

Catalog No. 5000204277
Encompass_Preferred

Amino-PEG10-OH is a non-cleavable, 10-unit polyethylene glycol (PEG) linker bearing a terminal amino group and a hydroxyl group. It is designed for bioconjugation applications, including the synthesis of antibody-drug conjugates and proteolysis-targeting chimeras (PROTACs), and is supplied for research use in small, controlled quantities.

  • Used in antibody-drug conjugate (ADC) and proteolysis-targeting chimera (PROTAC) synthesis.
  • Non-cleavable 10-unit PEG spacer with terminal amino and hydroxyl functionality.
  • Purity >98% as reported by manufacturer catalogs.
  • CAS registry number 129449-09-8 for unambiguous identification.
  • Molecular formula C20H43NO10 and molecular weight ~457.55 g/mol.
  • Appearance: colorless liquid or white solid, depending on lot and form.
  • Store at -20 °C for long-term storage; follow supplier safety data sheet for handling.

Catalog No. 50-002-04277 Supplier Medchemexpress LLC Supplier No. HY120761100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amino-PEG10-OH is a non-cleavable, 10-unit polyethylene glycol (PEG) linker bearing a terminal amino group and a hydroxyl group. It is designed for bioconjugation applications, including the synthesis of antibody-drug conjugates and proteolysis-targeting chimeras (PROTACs), and is supplied for research use in small, controlled quantities.

  • Used in antibody-drug conjugate (ADC) and proteolysis-targeting chimera (PROTAC) synthesis.
  • Non-cleavable 10-unit PEG spacer with terminal amino and hydroxyl functionality.
  • Purity >98% as reported by manufacturer catalogs.
  • CAS registry number 129449-09-8 for unambiguous identification.
  • Molecular formula C20H43NO10 and molecular weight ~457.55 g/mol.
  • Appearance: colorless liquid or white solid, depending on lot and form.
  • Store at -20 °C for long-term storage; follow supplier safety data sheet for handling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.