Learn More
Medchemexpress LLC 29-amino-3,6,9,12,15,18,21,24,27-nonaoxanonacosan-1-ol | 129449-09-8 | MFCD28385478 | >95.0% | 457.56 g/mol | C20H43NO10 | 1 G
SDP
Catalog No. 5000204278
Encompass_Preferred
Amino-PEG10-OH is a 10-unit polyethylene glycol (PEG) linker with a terminal amino group and a hydroxyl group, used as a non-cleavable, hydrophilic spacer in antibody-drug conjugate and PROTAC synthesis to improve solubility and control spacing between functional moieties.
  • Non-cleavable 10-unit PEG spacer suitable for ADC and PROTAC chemistry.
  • Terminal amino and hydroxyl groups enable versatile conjugation chemistries.
  • Enhances aqueous solubility of conjugates.
  • Provides a predictable linker length for spatial control.
  • Available in small-scale quantities for laboratory research.
Catalog No. 50-002-04278 Supplier Medchemexpress LLC Supplier No. HY1207611G
Amino-PEG10-OH is a 10-unit polyethylene glycol (PEG) linker with a terminal amino group and a hydroxyl group, used as a non-cleavable, hydrophilic spacer in antibody-drug conjugate and PROTAC synthesis to improve solubility and control spacing between functional moieties.
  • Non-cleavable 10-unit PEG spacer suitable for ADC and PROTAC chemistry.
  • Terminal amino and hydroxyl groups enable versatile conjugation chemistries.
  • Enhances aqueous solubility of conjugates.
  • Provides a predictable linker length for spatial control.
  • Available in small-scale quantities for laboratory research.