Learn More
Cellvis 29 MM GLASS BOTTOM DISH WITH 1
Catalog No. NC1335722
Encompass
29 mm Glass bottom dish with 10 mm micro-well #1.5 cover glass
Catalog No. NC1335722 Supplier Cellvis Supplier No. D29101.5N
29 mm Glass bottom dish with 10 mm micro-well #1.5 cover glass