Learn More
Cellvis 29 MM GLASS BOTTOM DISH WITH 1
Catalog No. NC0662883
Encompass
29mm glass bottom dish, dish size 29mm, well size 14mm, #1.5 glass(0.16-0.19mm).
Catalog No. NC0662883 Supplier Cellvis Supplier No. D29141.5N
29mm glass bottom dish, dish size 29mm, well size 14mm, #1.5 glass(0.16-0.19mm).